Learn More
Kautex™ LDPE Wide Mouth Bottles without Closures
Product Code. 10797551
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
300 mL
1500 mL
Unit Size:
Each
Pack of 360
Pack of 46
Product Code. Capacity (Metric) unitSize
10797551 50 mL Each
10129760 100 mL Each
15921391 100 mL Pack of 360
10116360 300 mL Each
15951391 1500 mL Pack of 46
5 options
Product Code. 10797551
Brand: Kautex™ 2000070528
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed with a wide opening for easy filling and cleaning. Kautex™ Wide Mouth Bottles are ideal for liquids, pastes and granules.
  • Manufactured from low density polyehtylene (LDPE) for flexibility and toughness
  • Natural translucent color, round bottles
  • Ideal for dispensing, sending samples, adhesives, transportation and storage 
  • Closures available separately
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
Form Wide Neck
Material LDPE
Quantity 1Each
Closure Included No
Graduated No
Shape Round
Product Type Wide Neck Bottle