Learn More
Lovibond™ Glass Filter Set for SpectroDirect & XD7000 Spectrophotometer
Product Code. 11715438
Change view
Click to view available options
Type:
Secondary Standard Set
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11715438 Secondary Standard Set Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11715438 Supplier Lovibond™ Supplier No. 711160
  • ONLINE EXCLUSIVE PRICE
    21180.00 NOK
    19464.15 NOK / Pack of 1
    Save 1715.85 NOK
    8% Off
Estimated Shipment: 23-11-2026
Complete Glass Filter Set for testing the photometric accuracy and the wavelength accuracy of the spectrophotometer
Specifications
Type Secondary Standard Set
For Use With (Equipment) SP600 spectrophotometer
Includes 4 Glass filters for wavelength confirmation