missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Replacement Reagents for CHECKIT™ Comparator: Silica No. 2 Tablets
Specifications
Specifications
| Color | Beige |
| Composition | (+)-Tartaric Acid 40 to 45%, Boric Acid 35 to 40%, non-hazardous components 25 to 35% |
| pH Range | 3.2 |
| Format | Tablet |
| For Use With (Application) | LR silica Aqua comparator™ test kit, silica VLR Aqua comparator™ test kit, silica reagent set, water analysis |
| For Use With (Equipment) | Silica SC450 colorimeter, MC500 colorimeter, SP600 spectrophotometer |
| Range | 0.05 to 4mg/L |
| Test Method | Silicomolybdate |
| Type | Silica No. 2 Tablet |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?