Learn More
Lovibond™ Total Chlorine Powder Packs™
Product Code. 15351655
Change view
Click to view available options
Quantity:
100 Pack
1000 Pack
Range:
0 to 2 mg/L
0 to 3.5 mg/L
0 to 5 mg/L
Volume (English):
0.16 oz.
0.33 oz.
0.84 oz.
Volume (Metric):
5 mL
10 mL
25 mL
Unit Size:
Pack of 100
Pack of 1000
Product Code. Quantity Range Volume (English) Volume (Metric) unitSize
15351655 100 Pack 0 to 5 mg/L 0.84 oz. 25 mL Pack of 100
15351645 100 Pack 0 to 3.5 mg/L 0.16 oz. 5 mL Pack of 100
15361645 1000 Pack 0 to 3.5 mg/L 0.16 oz. 5 mL Pack of 1000
11709039 1000 Pack 0 to 2 mg/L 0.33 oz. 10 mL Pack of 1000
15341655 1000 Pack 0 to 2 mg/L 0.33 oz. 10 mL Pack of 1000
11718999 100 Pack 0 to 2 mg/L 0.33 oz. 10 mL Pack of 100
10402345 100 Pack 0 to 2 mg/L 0.33 oz. 10 mL Pack of 100
15361655 1000 Pack 0 to 5 mg/L 0.84 oz. 25 mL Pack of 1000
11719039 1000 Pack 0 to 5 mg/L 0.84 oz. 25 mL Pack of 1000
9 options
Product Code. 15351655
Brand: Lovibond™ 530130
Premeasured unit dose DPD reagents for water quality testing
  • For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed foil packets eliminate risk of cross-contamination
  • Available for 5, 10, and 25mL sample sizes
  • Extended shelf life of up to 5 years
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted DPD method
Specifications
Certifications/Compliance US EPA
Volume (Metric) 25 mL
Format Powder
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer
Quantity 100 Pack
Range 0 to 5 mg/L
Type DPD Total Chlorine Reagent
Volume (English) 0.84 oz.