Learn More
Lovibond™ MC500 Colorimeter Replacement Reagents: Alkalinity-m (total)
For use with MC500 multi-parameter colorimeter
Brand: Lovibond™ 513210BT
Additional Details : Weight : 0.30000kg
Description
For use with MC500 multi-parameter colorimeter
- Parameter: Alkalinity-m (total)
- Method: Acid/Indicator
Specifications
Yellow | |
Tablet | |
MC500 multi-parameter colorimeter | |
5 to 300mg/L | |
Alka-M Tablet |
3.5 | |
Water quality testing | |
100 | |
Acid/Indicator |