Learn More
Lovibond™ Infrared Lab Turbidimeter
Product Code. 10247642
Change view
Click to view available options
Display Type:
Color Graphic Display
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Type Unit Size
10247642 Color Graphic Display Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10247642 Supplier Lovibond™ Supplier No. 194000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
All items necessary for testing including turbidimeter, four sample cells, calibration set, operator's manual, and carrying case. (Batteries sold separately.)
Designed for quick and easy turbidity measurement in the laboratory
  • Auto-ranging function automatically selects the correct range for turbidity measurement from 0.01 to 1100 NTU
  • Data-logging feature enables user to store up to 1,000 data sets with the date, time, and registration number
  • Instrument is power efficient; operates on seven NiCd rechargeable batteries (sold separately) or wall plug adapter
Specifications
Type Infrared Lab Turbidity Meter
Light Source Infrared LED
Display Type Color Graphic Display
Method Nephelometric
Sample Size 12 mL
For Use With (Application) For quick and easy turbidity measurement in the laboratory
Accuracy ±2%, ±5%
Resolution 0.01, 0.1, 1 NTU
Turbidity 0.01 to 1100 NTU
Dimensions (L x W x H) 10.45 x 7.6 x 2.76 in. (26.5 x 19.3 x 7 cm)
Length (English) 10.45 in.
Length (Metric) 26.5 cm
Width (English) 7.6 in.
Width (Metric) 19.3 cm
Height (English) 2.76 in.
Height (Metric) 7 cm
Battery Type NiCd rechargeable
Electrical Requirements 100/230 V 50/60 Hz
Frequency 50/60 Hz
Voltage 100/230 V
Certifications/Compliance CE, ISO
Temperature (English) Operating 41°F to 104°F
Temperature (Metric) Operating 5°C to 40°C
Weight (English) 2.2 lb.
Weight (Metric) 1 kg