Learn More
Lovibond™ Color of Varnish (Gardner Color) Discs
Brand: Lovibond™ 243030
Additional Details : Weight : 0.25000kg
Specifications
ASTM D-1544, AOCS Td 1a | |
Color of Varnish l Disc | |
Varnish comparator kits 705-V2, 705-VA2 or 705-VA4 | |
1 to 9 |