missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Aluminum
Click to view available options
Range:
0 to 0.3mg/L
Unit Size:
Each
Description
Includes
- Aqua Comparator Housing
- Aqua Comparator Disc
- Aluminum No. 1 Tablets (100 pack)
- Aluminum No. 2 Tablets (100 pack)
- 2 x 13.5mm Sample Cells
- Tablet Crusher
- Instructions
- Carrying Case
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
Specifications
| Accuracy | ±5% |
| For Use With (Application) | Routine water analysis |
| Includes | Aqua comparator™ housing, aqua comparator™ disc, aluminum No. 1 tablets, aluminum No. 2 tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case |
| No. of Tests | 100 |
| Range | 0 to 0.3mg/L |
| Type | Aluminum Test Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction