missing translation for 'onlineSavingsMsg'
Learn More

Loteprednol Etabonate, MedChemExpress

Product Code. p-200102056
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
16443317 50 mg 50mg
16417256 100 mg 100mg
16407308 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 16443317

Brand: Medchem Express HY17358

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Loteprednol etabonate (LE) is an orally active soft steroid belonging to a unique class of glucocorticoids. Loteprednol etabonate (LE) exhibits anti-inflammatory activity and has been used in optometry and ophthalmology.

Loteprednol etabonate (LE) is an orally active soft steroid belonging to a unique class of glucocorticoids. Loteprednol etabonate (LE) exhibits anti-inflammatory activity and has been used in optometry and ophthalmology.

Specifications

Chemical Name or Material Loteprednol Etabonate
CAS 82034-46-6
Color White
Purity Grade Notes Research
Molecular Formula C24H31ClO7
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (214.16 mM; Need ultrasonic)
SMILES O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])CC[C@]4(OC(OCC)=O)C(OCCl)=O)=C1
Molecular Weight (g/mol) 466.95
Formula Weight 466.95
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.