Learn More
Loteprednol Etabonate, MedChemExpress
Product Code. 16443317
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
16443317 50 mg 50mg
16417256 100 mg 100mg
16407308 10 mg 10mg
3 options
Product Code. 16443317
Brand: Medchem Express HY17358
MedChemExpress Loteprednol etabonate (LE) is an orally active soft steroid belonging to a unique class of glucocorticoids. Loteprednol etabonate (LE) exhibits anti-inflammatory activity and has been used in optometry and ophthalmology.
Loteprednol etabonate (LE) is an orally active soft steroid belonging to a unique class of glucocorticoids. Loteprednol etabonate (LE) exhibits anti-inflammatory activity and has been used in optometry and ophthalmology.
Specifications
Chemical Name or Material Loteprednol Etabonate
CAS 82034-46-6
Color White
Purity Grade Notes Research
Molecular Formula C24H31ClO7
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (214.16 mM; Need ultrasonic)
SMILES O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])CC[C@]4(OC(OCC)=O)C(OCCl)=O)=C1
Molecular Weight (g/mol) 466.95
Formula Weight 466.95
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation