Learn More
Lotamilast, MedChemExpress
Product Code. 16478017
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16478017
Brand: Medchem Express HY12740
MedChemExpress Lotamilast (RVT-501; E6005) is a selective phosphodiesterase 4 (PDE4) inhibitor with an IC50 of 2.8 nM.
Lotamilast (RVT-501
Specifications
Chemical Name or Material Lotamilast
CAS 947620-48-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H24N4O5
Quantity 5 mg
Synonym RVT-501 E6005
Solubility Information DMSO : 25 mg/mL (52.91 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C(OC)=O)C=C1)NC2=CC=CC(C3=C4C(C=C(OC)C(OC)=C4)=NC(NC)=N3)=C2
Molecular Weight (g/mol) 472.49
Formula Weight 472.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.