Learn More
Lorediplon, MedChemExpress
Product Code. 16453727
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16453727
Brand: Medchem Express HY19371
MedChemExpress Lorediplon is a novel non-benzodiazepine drug acting as a GABAA receptor modulator, differentially active at the alpha1-subunit, associated with promoting sleep.
Lorediplon is a novel non-benzodiazepine drug acting as a GABAA receptor modulator, differentially active at the alpha1-subunit, associated with promoting sleep.
Specifications
Chemical Name or Material Lorediplon
CAS 917393-39-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H15FN4O2S
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (253.54 mM; Need ultrasonic)
SMILES CC(N(C1=CC(C2=CC=NC3=C(C(C4=CC=CS4)=O)C=NN23)=CC=C1F)C)=O
Molecular Weight (g/mol) 394.42
Formula Weight 394.42
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.