Learn More
SeaKem™ GTG™ Agarose, Lonza™
Product Code. p-2861586
Click to view available options
Quantity:
25 g
500 g
Unit Size:
25g
500g
Product Code. 11540727
Brand: Lonza 50071
Specifically designed for preparative DNA electrophoresis. Lonza™ SeaKem™ GTG™ Agarose is a standard gelling temperature, high gel strength agarose that resolves DNA fragments greater than 1,000 bp.
  • DNA recovered from SeaKem™ GTG™ Agarose gels may be reliably digested and ligated
  • Extensively performance tested to ensure complete compatibility with routine molecular biology techniques
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material SeaKem GTG Agarose
Quantity 25 g