Learn More
Lonza™ Reliant™ UV Transparent Tray
Landscape format, open ended tray designed to give you clear, straight, band patterns
Brand: Lonza™ 54945
Additional Details : Weight : 0.45000kg
Description
Remove tape from base of tray before use.
Specifications
UV Transparent | |
Electrophoresis System | |
Reliant™ Gel System Tray | |
Acrylic |