Learn More
Lonza™ IsoGel™ IEF Plates
Product Code. 11517995
Change view
Click to view available options
pH Range:
3 to 10
6 to 10.5
7 to 11
Unit Size:
Each
Pack of 6
Product Code. pH Range unitSize
11517995 6 to 10.5 Pack of 6
11507995 3 to 10 Pack of 6
12890125 7 to 11 Each
3 options
Product Code. 11517995
Brand: Lonza™ 56018
Precast agarose IEF plates provide rapid and reproducible isoelectric focusing of proteins up to 2000kDa
  • Specifically designed for antibody analysis of
  • Each gel is supported on GelBond™ film for easy handling and dimensional stability
  • Gel size is compatible with most IEF chambers
  • 125 x 100mm
Isoelectric focusing, antibody separation and analysis
Order Info
Accessories packs (Cat. No. BMA56014 and BMA56010) and blotting paper (BMA56007) available separately.
TRUSTED_SUSTAINABILITY
Specifications
pH Range 6 to 10.5
Separation Type Isoelectric Focusing
Form Solid
For Use With (Application) Isoelectric focusing, Antibody separation and analysis
Storage Requirements 2° to 8°C
Product Type Gel Plate