missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ DNA Ladders
Each band has 50% GC content for precise sizing and sharp resolution
Brand: Lonza™ 50321
Additional Details : Weight : 1.00000kg
Specifications
100 ng/μL | |
Liquid | |
Size determination of DNA samples | |
10 | |
2° to 8°C |
DNA Molecular Weight Marker/Ladder | |
Supplied at various concentrations in TE Buffer and each includes a vial of 6X Sample Loading Buffer 1mL total volume 15% w/v Ficoll 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene cyanol FF, 30mM EDTA | |
100bp | |
160 μL |