missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ DNA Ladders

Each band has 50% GC content for precise sizing and sharp resolution

Brand:  Lonza™ 50320

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11560777

  • 3270.00 NOK / 100 reactions
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

200 ng/μL
Liquid
Size determination of DNA samples
50
2° to 8°C
DNA Molecular Weight Marker/Ladder
Supplied at various concentrations in TE Buffer and each includes a vial of 6X Sample Loading Buffer 1mL total volume 15% w/v Ficoll 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene cyanol FF, 30mM EDTA
20bp
150 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers