missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ DNA Ladders
Click to view available options
Quantity:
150 μL
160 μL
300 μL
No. of Bands:
10
16
25
30
Concentration:
100 ng/μL
200 ng/μL
Increments:
100bp
20bp
500bp
Unit Size:
100 reactions
Each
Specifications
Specifications
| Concentration | 100 ng/μL |
| Product Type | DNA Molecular Weight Marker/Ladder |
| Form | Liquid |
| Formulation | Supplied at various concentrations in TE Buffer and each includes a vial of 6X Sample Loading Buffer 1mL total volume 15% w/v Ficoll 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene cyanol FF, 30mM EDTA |
| For Use With (Application) | Size determination of DNA samples |
| Increments | 100bp |
| No. of Bands | 10 |
| Quantity | 160 μL |
| Storage Requirements | 2° to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction