Learn More
Lonza™ BioWhittaker™ ProFreeze™ NAO Freezing Medium
Brand: Lonza™ 12-769E
Additional Details : Weight : 0.90720kg
Description
ProFreeze™-CDM, NAO, Chemically Defined Freeze Medium (2X) is universally suitable for cryopreserving many cell types in the absence of fetal bovine serum (FBS). However, it is used to greatest advantage with cells cultured in a serum-free and animal component-free environment. This protein-free freezing medium contains no animal derived components, insulin, or hydrolysate, and maintains high cell viability upon recovery from frozen storage.
Specifications
2° to 8°C | |
Liquid | |
100 mL |
2 X | |
Freezing Medium |