Learn More
Lonza™ BioWhittaker Insect-XPRESS Insect Cell Culture Media (1X) with L-Glutamine
Designed to support the growth of insect cell lines derived from Spodoptera frugiperda (Sf9 and Sf21). BioWhittaker™ Insect-XPRESS™ Insect Cell Culture Media (1X) with L-Glutamine supports superior production of recombinant proteins when using the Baculovirus Expression Vector System (BEVS).
Brand: Lonza™ BE12-730F
Additional Details : Weight : 1.36080kg
Description
- Protein-free
- Cell densities in excess of 8.3×106 cells/mL can be achieved with suspension cultures of Sf9 cells using Insect-XPRESS Medium and excess of oxygen
- Formulation can also be used for stationary mono¬layer cultures and shake-flask cultures
Specifications
2° to 8°C | |
Insect Cell Medium, Protein-free | |
500 mL |
Liquid | |
Glutamine |