Learn More
Lonza™ BioWhittaker™ Antibiotics: Penicillin-Streptomycin Mixtures
Used as a broad spectrum bacteriostatic and bactericidal, with activity against gram negative and gram-positive organisms including yeast and mold. BioWhittaker™ Antibiotics: Penicillin-Streptomycin Mixtures are manufactured in accordance with cGMP regulations.
Brand: Lonza™ DE17-603E
Additional Details : Weight : 0.01000kg
Description
- Suggested working concentration of Penicillin-Streptomycin is 100 units of Potassium Penicillin and 100 μg of Streptomycin Sulfate per 1 ml of culture media
- Manufactured in accordance with cGMP regulations
- Cell culture tested to assure compatibility.
- 20mL vials have crimp-top closure
- 4.5, 100 and 500mL bottles have screw-cap closure
Specifications
-10° to -20°C | |
5K/5K | |
100 mL |
Antibiotic | |
3.61 to 7.29 |
Safety and Handling
- Penicillin-Streptomycin
Hazard Statement
- EUH210-Safety data sheet available on request.
- EUH208-Contains Penicillin-G potassium salt. May produce an allergic reaction.
Supplemental information
- MIXTURE LIST-Contains : Streptomycin sulphate, Penicillin-G potassium salt