Learn More
Lonza™ AccuGENE™ 3M Sodium Acetate, pH 5.2
Product Code. 11578015
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11578015 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11578015 Supplier Lonza™ Supplier No. 51203
Ideal for a wide range of molecular biology applications
  • Ready-to-use
  • Prepared with 18megohm water
  • Filtered using 0.2μ filter and filled into sterile bottles
  • Manufactured according to strict quality control standards to ensure consistency
  • Guaranteed DNase, RNase and protease-free
  • Customized solutions are available to meet individual needs
TRUSTED_SUSTAINABILITY
Specifications
Concentration 3 M
DNase Yes
For Use With (Application) Ideal for a wide range of molecular biology applications
pH 5.2
Product Type 3M Sodium Acetate
RNase Yes
Quantity 500 mL
For research or manufacturing use only.