Learn More
Abnova™ LongTaq DNA Polymerase
Description
- The enzyme has been proved not having the 3′-5′ exonuclease activity.
- The enzyme has proven to have high amplification yield, be stable at high temparetaure.
Specifications
Specifications
| Concentration | 5 U/μL |
| Content And Storage | Store at -20°C Guaranteed stable for 6 months when properly stored |
| Quantity | 2500 units |
| For Use With (Application) | Standard PCR |
| Storage Buffer | 20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol |