Learn More
Abnova™ LongTaq DNA Polymerase
Product Code. 16052555
Click to view available options
Quantity:
1000 units
2500 units
500 units
Unit Size:
1000 units
2500 units
500 units
Product Code. 16052555
Brand: Abnova™ D0013.1000Units
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LongTaq DNA polymerase catalyzes 5'-3' synthesis of DNA.
  • The enzyme has been proved not having the 3′-5′ exonuclease activity.
  • The enzyme has proven to have high amplification yield, be stable at high temparetaure.
Specifications
Concentration 5 U/μL
Content And Storage Store at -20°C Guaranteed stable for 6 months when properly stored
Quantity 1000 units
For Use With (Application) Standard PCR
Storage Buffer 20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol