Learn More
Hamilton™ Long Life Instrument Syringes
Description
- Plunger tip made from UHMWPE (Ultra High Molecular Weight Poly Ethylene) which is harder than PTFE to better resist scratching
- Plunger rod made from 316 stainless steel which offers exceptional chemical resistance
Specifications
Specifications
| Material | Borosilicate Glass, PTFE, UHMWPE |
| Diameter (Metric) Outer | 7.73 mm |
| Product Type | Instrument Syringe |
| Quantity | 1 Ea. |
| Volume (Metric) | 100 μL |
| Connection Type | Luer Lock |