Learn More
Hirschmann™ Long Conical Centrifuge Tubes
Click to view available options
Graduations:
0 to 10 mL
Unit Size:
Pack of 100
Description
- Long conical form
- Available with either white or no graduations
Specifications
Specifications
| Capacity (Metric) | 15 mL |
| No. per Pack | 100 |
| Length (Metric) | 115 ±5 mm |
| Graduations | 0 to 10 mL |
| Product Type | Centrifuge Tube |