Learn More
Lomerizine Dihydrochloride, TRC
Product Code. 30137753
Change view
Click to view available options
Quantity:
10 mg
250 mg
500 mg
Unit Size:
10mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137753 250 mg 250mg
30133933 500 mg 500mg
30005598 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137753 Supplier Toronto Research Chemicals Supplier No. TRCL469430250MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101477-54-7
Molecular Formula C27 H30 F2 N2 O3 . 2 Cl H
Molecular Weight (g/mol) 541.46
Synonym Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-[(2,3,4-trimethoxyphenyl)methyl]-, hydrochloride (1:2) (ACI), Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-[(2,3,4-trimethoxyphenyl)methyl]-, dihydrochloride (9CI), KB 2796, Lomerizine dihydrochloride, Lomerizine hydrochloride, 1-[Bis(4-fluorophenyl)methyl]-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine hydrochloride (1:2)
IUPAC Name 1-[bis(4-fluorophenyl)methyl]-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine;dihydrochloride
SMILES Cl.Cl.COc1ccc(CN2CCN(CC2)C(c3ccc(F)cc3)c4ccc(F)cc4)c(OC)c1OC
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C27H30F2N2O3.2ClH/c1-32-24-13-8-21(26(33-2)27(24)34-3)18-30-14-16-31(17-15-30)25(19-4-9-22(28)10-5-19)20-6-11-23(29)12-7-20;;/h4-13,25H,14-18H2,1-3H3;2*1H
Formula Weight 540.1758
Chemical Name or Material Lomerizine, Dihydrochloride
Product Title
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.