Learn More
Lodelaben, MedChemExpress
Product Code. 16495607
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495607 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16495607 Supplier Medchem Express Supplier No. HY100240
MedChemExpress Lodelaben is a human neutrophil elastase inhibitor with an IC50 and Ki of 0.5 and 1.5 μM, respectively.
Lodelaben is a human neutrophil elastase inhibitor with an IC50 and Ki of 0.5 and 1.5 μM, respectively.
Specifications
Chemical Name or Material Lodelaben
Physical Form Solid
CAS 111149-90-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H41ClO3
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym SC-39026 Declaben
SMILES O=C(O)C1=CC=C(C(O)CCCCCCCCCCCCCCCCC)C=C1Cl
Molecular Weight (g/mol) 425.04
Formula Weight 425.04
Percent Purity 95.0%
Grade Research