Learn More
Brady™ Lock-in Chain
Product Code. 11721645 Shop All Brady Products
Change view
Click to view available options
Length (Metric):
6.1 m
Unit Size:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11721645 6.1 m 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11721645 Supplier Brady™ Supplier No. 077207
  • ONLINE EXCLUSIVE PRICE
    700.00 NOK
    686.26 NOK / Pack of 1
    Save 13.74 NOK
    1% Off
Estimated Shipment: 30-11-2026
Lock-in Chain are a quick, easy way to create a pedestrian barricade with the BradyCone™ Warning System.
  • Chains are made of yellow plastic links and lock directly into cone.
Specifications
Product Type Lock-in Chain
Color Yellow
For Use With (Application) Traffic Control
Length (English) 20 ft.
Length (Metric) 6.1 m
Quantity 1 Ea.