Learn More
Heinz Herenz LMS-Tube LIM Broth
Description
Our transport and collection swabs contain special applicators and test tubes with different liquid media, customized for the transportation and preservation of microbiological samples.
- The liquid medium allows the division of the sample material and thus several tests from one sample.
- The ready to use devices are supplied in a peelpack which is easy to open.
- The breakpoint in the shaft of the applicator enables a safe and hygienic workflow.
- The standard size of the test tubes with screwcap simplified the use with automatic analyzers.
- There are labels on the test tubes for a accurate sample identification.
- The items are produced according to the conditions of GMP (Good Manufacturing Practice) and certified according to DIN EN ISO 13485. This ensures a high quality.
- Our swabs are sterile (SAL 106), pyrogen free and noncytotoxic.
Specifications
Specifications
| Product Type | Transport and Collection Swab |
| Format | Swab |
| Sterility | Sterile |
| Color | White |
Product Title
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?