missing translation for 'onlineSavingsMsg'
Learn More

LMP744 hydrochloride, MedChemExpress

Product Code. 16433269
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16433269 1 mg 1mg
16482478 5 mg 5mg
16470279 10 mg 10mg
16455437 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16433269 Supplier Medchem Express Supplier No. HYU00248A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LMP744 hydrochloride (MJ-III65 hydrochloride) is a DNA intercalator and Topoisomerase I (Top1) inhibitor with antitumor activity.

LMP744 hydrochloride (MJ-III65 hydrochloride) is a DNA intercalator and Topoisomerase I (Top1) inhibitor with antitumor activity.

Specifications

Chemical Name or Material LMP744 hydrochloride
CAS 308246-57-3
Color Dark Brown
Purity Grade Notes Research
Molecular Formula C24H25ClN2O7
Quantity 1 mg
Synonym MJ-III65 hydrochlorideNSC706744 hydrochloride
Solubility Information DMSO : 105 mg/mL (214.76 mM; Need ultrasonic)
SMILES OCCNCCCN1C2=C(C(C3=CC(OCO4)=C4C=C23)=O)C5=CC(OC)=C(OC)C=C5C1=O.Cl
Molecular Weight (g/mol) 488.92
Formula Weight 488.92
Percent Purity 98.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.