missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMBRD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27203-1-AP-20UL
This item is not returnable.
View return policy
Description
LMBRD1, also known as NESI (nuclear export signal-interacting protein, is a lysosomal membrane protein that is thought be involved in the transport and metabolism of cobalamin. LMBRD1 was initially identified as interacting with the large form of the hepatitis delta antigen and may be required for the nucleocytoplasmic shuttling of the hepatitis delta virus. Mutations in this gene are associated with the vitamin B12 metabolism disorder termed, homocystinuria-megaloblastic anemia complementation type F (cblF).Specifications
| LMBRD1 | |
| Polyclonal | |
| Unconjugated | |
| LMBRD1 | |
| C6orf209, cblF, CD001, LMBD1, LMBR1 domain containing 1, LMBRD1, MSTP044, NESI, RP11 810I22.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55788, 68421 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8K0B2, Q9NUN5 | |
| LMBRD1 | |
| LMBRD1 Fusion Protein Ag24478 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction