missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LL-37 Antibody (B-T37) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18120JF635
This item is not returnable.
View return policy
Description
LL-37 Monoclonal antibody specifically detects LL-37 in Human samples. It is validated for Flow Cytometry,ELISA
Specifications
| LL-37 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| 18 kDa cationic antimicrobial protein, Antibacterial peptide FALL-39, CAP18, CAP-18, cathelicidin antimicrobial peptide, CRAMP, epididymis secretory sperm binding protein, FALL39, FALL-39, FALL-39 peptide antibiotic, hCAP-181, HSD26, LL37 | |
| Human antimicrobial peptide LL-37. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| 0.1 mL | |
| Innate Immunity | |
| 820 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA | |
| B-T37 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction