Learn More
Lithium tartrate monohydrate, 99%
Product Code. p-7050730
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378118 50 g 50g
11388118 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11378118 Supplier Thermo Scientific Alfa Aesar Supplier No. 039357.18
Request Bulk or Custom Format
CAS: 6108-32-3 | C4H6Li2O7 | 179.965 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6108-32-3
Molecular Formula C4H6Li2O7
Molecular Weight (g/mol) 179.965
MDL Number MFCD00054421
InChI Key GNDOSKKVQUGWFB-UHFFFAOYSA-L
Synonym lithium tartrate monohydrate, lithium tartrate 1-hydrate, c4h4o6.2li.h2o, dilithium 1+ hydrate tartrate, dilithium 1+ ion hydrate tartrate, lithium 2,3-dihydroxybutanedioate-water 2/1/1
PubChem CID 18463206
IUPAC Name dilithium;2,3-dihydroxybutanedioate;hydrate
SMILES [Li+].[Li+].C(C(C(=O)[O-])O)(C(=O)[O-])O.O
Specifications
Odor Odorless
Quantity 50 g
Solubility Information Soluble in water
Formula Weight 179.97 (161.97 Anhydrous)
Percent Purity 99%
Physical Form Powder or Aggregates
Chemical Name or Material Lithium tartrate monohydrate