Learn More
Lithium tantalum oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog Number 11324797
Click to view available options
Quantity:
10 g
50 g
12031-66-2
LiO3Ta
235.88
MFCD00016174
JNQQEOHHHGGZCY-UHFFFAOYSA-N
lithium tantalate, lithium oxido dioxo tantalum, lithium 1+ tantalumoylolate
5148101
lithium;oxido(dioxo)tantalum
[Li+].[O--].[O--].[O--].[Ta+5]
12031-66-2
LiO3Ta
235.88
MFCD00016174
JNQQEOHHHGGZCY-UHFFFAOYSA-N
lithium tantalate, lithium oxido dioxo tantalum, lithium 1+ tantalumoylolate
5148101
lithium;oxido(dioxo)tantalum
[Li+].[O--].[O--].[O--].[Ta+5]
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 10 g
Formula Weight 235.89
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Lithium tantalum oxide