Learn More
Lithium Sulfate Monohydrate, Supelco™
Product Code. 15867380
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15867380 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
Product Code. 15867380 Supplier Supelco Supplier No. 1056940250
 Restricted Item
Estimated Shipment: 26-11-2026
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
For analysis EMSURE™ ACS,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 120°C
CAS 10102-25-7
Quantity 250 g
MDL Number MFCD00149766
Synonym Lithium sulfate monohydrate, Lithium sulfate monohydrate
Molecular Weight (g/mol) 109.94 (Anhydrous Basis)
Grade Research
Density 2.05 g/cm3 at 20°C
Chemical Name or Material Lithium Sulfate Monohydrate