Learn More
Lithium sulfate, anhydrous, TRC
Product Code. 30468962
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468962 50 g 50g
30475976 10 g 10g
30479295 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30468962 Supplier Toronto Research Chemicals Supplier No. TRCL46937350G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10377-48-7
Molecular Formula Li2SO4
Molecular Weight (g/mol) 109.95
IUPAC Name dilithium;sulfate
SMILES [Li+].[Li+].[O-]S(=O)(=O)[O-]
Specifications
Quantity 50 g
InChI Formula InChI=1S/2Li.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
Formula Weight 109.9837
Chemical Name or Material Lithium sulfate, anhydrous