missing translation for 'onlineSavingsMsg'
Learn More

Lithium, Sigma-Aldrich™

Product Code. 15880231
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15880231 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15880231 Supplier Sigma Aldrich Supplier No. 8180440050
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

Granular (protective medium: argon for synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 7439-93-2
Melting Point 179°C
Density 0.534 g/cm3 at 20°C
Quantity 50 g
Form Granular
MDL Number MFCD00134051
Synonym Lithium, Lithium
Molecular Weight (g/mol) 6.94
Chemical Name or Material Lithium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.