missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Phosphate (Meta), MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| CAS | 13762-75-9 |
| Quantity | 100 g |
| Molecular Formula | LiO3P |
| Synonym | lithium metaphosphate, lithium polymetaphosphate, metaphosphoric acid, lithium salt, lithotab o3p, li.o3p, lithium phosphate meta, lithium 1+ ion trioxidophosphate 1- |
| InChI Key | MRVHOJHOBHYHQL-UHFFFAOYSA-M |
| SMILES | [Li+].[O-]P(=O)=O |
| Molecular Weight (g/mol) | 85.911 |
| PubChem CID | 23667996 |
| Formula Weight | 115.79 |
| Chemical Name or Material | Lithium Phosphate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction