Learn More
Lithium Carbonate, Supelco™
Product Code. 15837380
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15837380 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
Product Code. 15837380 Supplier Supelco Supplier No. 1056800250
 Restricted Item
Estimated Shipment: 26-11-2026
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
For analysis EMSURE™ ACS,Reag. Ph Eur
We are committed to bringing you Greener Alternative Products, which belong to one of the four categories of greener alternatives. LITHIUM CARBONATE is essential for sustainable energy solutions, particularly in lithium-ion batteries. Its eco-friendly properties enhance battery performance and longevity, supporting the transition to cleaner energy technologies while reducing environmental impact.
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 720°C
CAS 554-13-2
Quantity 250 g
MDL Number MFCD00011084
Synonym Lithium carbonate, Lithium carbonate
Molecular Weight (g/mol) 73.89
Grade Research
Density 2.1 g/cm3 at 20°C
Chemical Name or Material Lithium Carbonate