missing translation for 'onlineSavingsMsg'
Learn More

LIS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16850255 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16850255 20 μL 20µL
16840255 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16850255

Brand: Proteintech 206781AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VAMP7 is a member of the soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) family, localizing to late endosomes and lysosomes. VAMP7 is thought to mediate the fusion of endosomes to their target lysosomes as well as other exocytosis events during phagocytosis and neuritogenesis. VAMP7 interacts with the VPS9 ankyrin repeat protein VARP, a protein that localizes to early endosomes and thought to regulate endosome dynamics. Together with CD82, VAMP7 can modulate the signaling of EGFR by regulating its endocytosis from the plasma membrane.

Specifications

Antigen LIS1
Applications Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PAFAH1B1
Gene Accession No. P43034, P63004, P63005
Gene Alias LIS 1, LIS1, LIS2, Lissencephaly 1 protein, MDCR, MDS, PAF AH 45 kDa subunit, PAF AH alpha, PAFAH, PAFAH alpha, PAFAH1B1, PAFAHA
Gene Symbols Pafah1b1
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18472, 5048, 83572
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.