Learn More
Liquiritin, TRC
Product Code. 30135911
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135911 100 mg 100mg
30132768 25 mg 25mg
30007003 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135911 Supplier Toronto Research Chemicals Supplier No. TRCL468830100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 551-15-5
Molecular Formula C21 H22 O9
Molecular Weight (g/mol) 418.39
Synonym 4H-1-Benzopyran-4-one, 2-[4-(β-D-glucopyranosyloxy)phenyl]-2,3-dihydro-7-hydroxy-, (2S)-, 4H-1-Benzopyran-4-one, 2-[4-(β-D-glucopyranosyloxy)phenyl]-2,3-dihydro-7-hydroxy-, (S)-, Liquiritin (6CI,7CI,8CI), (2S)-2-[4-(β-D-Glucopyranosyloxy)phenyl]-2,3-dihydro-7-hydroxy-4H-1-benzopyran-4-one, 4',7-Dihydroxyflavanone 4'-β-D-glucopyranoside, 4',7-Dihydroxyflavanone 4'-β-D-glucoside, 7-Hydroxyflavanone 4'-O-glucoside, Likviritin, Liquiritigenin 4'-β-D-glucopyranoside, Liquiritoside
IUPAC Name (2S)-7-hydroxy-2-[4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]-2,3-dihydrochromen-4-one
SMILES OC[C@H]1O[C@@H](Oc2ccc(cc2)[C@@H]3CC(=O)c4ccc(O)cc4O3)[C@H](O)[C@@H](O)[C@@H]1O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C21H22O9/c22-9-17-18(25)19(26)20(27)21(30-17)28-12-4-1-10(2-5-12)15-8-14(24)13-6-3-11(23)7-16(13)29-15/h1-7,15,17-23,25-27H,8-9H2/t15-,17+,18+,19-,20+,21+/m0/s1
Formula Weight 418.1264
Chemical Name or Material Liquiritin