missing translation for 'onlineSavingsMsg'
Learn More

Liquiritin, MedChemExpress

Product Code. p-200096309
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16481388

Brand: Medchem Express HYN0376

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Liquiritin, a flavonoid isolated from Glycyrrhiza, is a potent and competitive AKR1C1 inhibitor with IC50s of 0.62 μM, 0.61 μM, and 3.72μM for AKR1C1, AKR1C2 and AKR1C3, respectively. Liquiritin efficiently inhibits progesterone metabolism mediated by AKR1C1 in vivo. Liquiritin acts as an antioxidant and has neuroprotective, anti-cancer and anti-inflammatory activity.

Liquiritin, a flavonoid isolated from Glycyrrhiza, is a potent and competitive AKR1C1 inhibitor with IC50s of 0.62 μM, 0.61 μM, and 3.72μM for AKR1C1, AKR1C2 and AKR1C3, respectively. Liquiritin efficiently inhibits progesterone metabolism mediated by AKR1C1 in vivo. Liquiritin acts as an antioxidant and has neuroprotective, anti-cancer and anti-inflammatory activity.

Specifications

Chemical Name or Material Liquiritin
CAS 551-15-5
Color White
Purity Grade Notes Research
Molecular Formula C21H22O9
Quantity 5 mg
Solubility Information DMSO : 150 mg/mL (358.52 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C1C[C@@H](C2=CC=C(O[C@H]3[C@@H]([C@H]([C@@H]([C@@H](CO)O3)O)O)O)C=C2)OC4=CC(O)=CC=C14
Molecular Weight (g/mol) 418.39
Formula Weight 418.39
Percent Purity 98.3%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.