Learn More
DWK Life Sciences Wheaton™ Liquid Scintillation Vials, PET
Shop All Wheaton Science Products ProductsDescription
Provides the clarity of glass with the safety of plastic.
- Vials offer low permeability to solvents and minimal background counts
- Clarity of glass with the safety of plastic
- Vials can be safely incinerated
- No harmful gas is generated
- Packaged in 5 utility trays with each partition tray holding 100 vials
- Utility trays serve as a way to store your samples
Specifications
Specifications
| Material (Liner) | Polyethylene Cone |
| Color | Clear |
| Diameter (Metric) Outer | 27 mm |
| Material | PET |
| Volume (Metric) | 20 mL |
| No. per Case | 1000 |
| Closure Material | Urea |
| Closure Size | 22-400 |
| Height (Metric) | 61 mm |