missing translation for 'onlineSavingsMsg'
Learn More

Liqui-Gel 29:1, MP Biomedicals

Product Code. 11441582 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441582 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11441582 Supplier MP Biomedicals Supplier No. 04800803

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A 40% (w/v) solution of Ultra Pure Acrylamide (38.67%) and bis-Acrylamide (1.33%) in specially-prepared deionized water. Final ratio is 29:1

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Liqui-Gel™ 29:1
Quantity 500 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Liqui-Gel 29:1
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.