Learn More
Invitrogen™ eBioscience™ Lipopolysaccharide (LPS) Solution (500X)
Description
- A ready to use solution of LPS from Escherichia coli 026:B6 in aqueous solution
- LPS is a major component of the cell wall of gram negative bacteria
- It is highly immunogenic and strongly activates immune cells bearing the CD14/TLR4/MD2 receptor complex
- This reagent is intended for use in in vitro culture of B cells, monocytes, macrophages and other cells that are responsive to LPS
Specifications
Specifications
| Cell Type | B cells, monocytes, macrophages and other cells that are responsive to LPS. |
| Concentration | 500 X |
| Form | Liquid |
| Product Type | LPS Solution |
| Sterility | Sterile |
| Product Line | eBioscience |
| Quantity | 4 x 100 μL |