missing translation for 'onlineSavingsMsg'
Learn More

LIPH Mouse anti-Human, Porcine, Clone: 1F5D10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66303-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16848055

  • 3828.53 NOK / 150µL
Estimated Shipment: 17-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a membrane-bound member of the mammalian triglyceride lipase family. It catalyzes the production of 2-acyl lysophosphatidic acid (LPA), which is a lipid mediator with diverse biological properties that include platelet aggregation, smooth muscle contraction, and stimulation of cell proliferation and motility.
Specifications

Specifications

LIPH
Monoclonal
2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WWY8
LIPH
LIPH Fusion Protein Ag24009
150 μL
Primary
Human, Porcine
Antibody
IgG2a
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1F5D10
Unconjugated
LIPH
AH, Lipase member H, lipase, member H, LIPH, LPD lipase related protein, LPDLR, mPA PLA1, mPA PLA1 alpha, MPAPLA1, Phospholipase A1 member B, PLA1B
Mouse
Protein A
RUO
100737989, 200879
-20°C
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers