Learn More
Linperlisib, MedChemExpress
Product Code. 16493688
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493688 10 mg 10mg
16486427 5 mg 5mg
16404776 100 mg 100mg
16400087 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16493688 Supplier Medchem Express Supplier No. HY102031
MedChemExpress Linperlisib (YY-20394) is a potent, orally bioavailable and selective inhibitor of PI3Kδ extracted from patent WO 2015055071 A1, compound 10; has an IC50 of 6.4 nM.
Linperlisib (YY-20394) is a potent, orally bioavailable and selective inhibitor of PI3Kδ extracted from patent WO 2015055071 A1, compound 10
Specifications
Chemical Name or Material Linperlisib
CAS 1702816-75-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H37FN6O5S
Quantity 10 mg
Synonym YY-20394
Solubility Information DMSO : 100 mg/mL (169.87 mM; Need ultrasonic)
SMILES CS(=O)(NC1=CC(C2=C3C=C(F)C=C(CN4CCC(C(C)(O)C)CC4)C3=NC(N5CCOCC5)=N2)=CN=C1OC)=O
Molecular Weight (g/mol) 588.69
Formula Weight 588.69
Percent Purity 98.95%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease