Learn More
MP Biomedicals™ Linbro™ 0.6 mL Snap-Top Microcentrifuge Tubes
Linbro™ 0.6 mL Snap-Top Microcentrifuge Tubes with reference lines for quick volume estimations and a frosted writing area.
Brand: MP Biomedicals™ 0219351990
Additional Details : Weight : 0.01000kg
Description
- Capacity: 0.6 mL
- Snap-top
- Features reference lines for quick volume estimations and a frosted writing area
Specifications
0.6 mL | |
Orange | |
Plastic | |
Snap Cap |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.