Learn More
DWK Life Sciences Limited Volume Insert
Shop All DWK Life Sciences Products
Click to view available options
Volume (Metric):
0.1 mL
Unit Size:
Pack of 100
Description
The DWK Life Sciences line of chromatography vials offers two different LVI styles: flat bottom and conical.
- Packed in zip-lock PE-bags with tear-off foil strip to ensure tamper-evidence and re-closeability
Specifications
Specifications
| Diameter (Metric) Outer | 6 mm |
| Material | Borosilicate Glass (5.1) |
| Volume (Metric) | 0.1 mL |
| For Use With (Application) | High Performance Liquid Chromatography (HPLC), Gas Chromatography (GC), Small Sample Volume Analysis |
| Base Style | Conical |
| Autoclavable | Autoclavable |
| Height (Metric) | 31 mm |
| Quantity | 100 |
| Thickness (Metric) | 0.7 mm |